Assay Details
| Gene Symbol: Dcun1d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163703.1 | NP_001157175.1 | 3 | 3929 | 75 | |
| NM_173408.3 | NP_775584.1 | 3 | 3948 | 75 | ||
| GenBank mRNA | AK006140.1 | - | 1 | 349 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 2610020H08 gene | 2610020H08Rik | Anti-sense | Chr. 7 - 119794130 - 119882695 on NCBI Build GRCm38 | ||
| Downstream | LYR motif containing 1 | Lyrm1 |
1110065L10Rik, 2310004B22Rik, 4930404J24Rik |
40.6kb | Anti-sense | Chr. 7 - 119895807 - 119956203 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dcun1d3
Genomic Map