Assay Details
| Gene Symbol: 1700047O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006720.1 | - | 1 | 454 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
vomeronasal 2, receptor 56 |
Vmn2r56 | EG629079 | -47.5kb | Anti-sense | Chr. 7 - 12693860 - 12733320 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 329 | Zfp329 |
2810439M05Rik, 4632409L22Rik, ZNF329 |
22.2kb | Anti-sense | Chr. 7 - 12803778 - 12818895 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700047O18Rik
Genomic Map