Assay Details
| Gene Symbol: 1700061D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006849.1 | - | 1 | 350 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA D830014E11 gene | D830014E11Rik | -33.9kb | Anti-sense | Chr. 5 - 105827095 - 105832437 on NCBI Build GRCm38 | |
| Downstream | zinc finger protein 326 | Zfp326 |
5730470H14Rik, ZAN75, Znf326 |
9.8kb | Anti-sense | Chr. 5 - 105876568 - 105915818 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700061D13Rik
Genomic Map