Assay Details
| Gene Symbol: Umps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007077.1 | - | 1 - 2 | 102 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | integrin beta 5 | Itgb5 |
AA475909, AI874634, ESTM23, [b]-5, [b]5, [b]5A, [b]5B, beta-5, beta5 |
-16.9kb | Sense | Chr. 16 - 33829665 - 33949338 on NCBI Build GRCm38 |
| Downstream |
kalirin, RhoGEF kinase |
Kalrn |
2210407G14Rik, AV235988, DUET, E530005C20Rik, Gm539, Hapip, TRAD |
1.8kb | Anti-sense | Chr. 16 - 33969073 - 34573291 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Umps
Genomic Map