Assay Details
| Gene Symbol: 1700121I08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007225.1 | - | 1 | 146 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5617 | Gm5617 | EG434402 | -4.0 | Anti-sense | Chr. 9 - 48495343 - 48495975 on NCBI Build GRCm38 |
| Downstream | nicotinamide N-methyltransferase | Nnmt | 41.6kb | Sense | Chr. 9 - 48538001 - 48607635 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700121I08Rik
Genomic Map