Assay Details
| Gene Symbol: 1810059C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007903.1 | - | 1 - 2 | 88 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 10 |
Mllt10 |
Af10, B130021D15Rik, D630001B22Rik, mKIAA4140 |
Anti-sense | Chr. 2 - 18055175 - 18212390 on NCBI Build GRCm38 | |
| Downstream | microRNA 7655 | Mir7655 |
Gm27794, mmu-mir-7655 |
1.2kb | Anti-sense | Chr. 2 - 18057846 - 18057903 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1810059C17Rik
Genomic Map