Assay Details
| Gene Symbol: 2210009P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008691.1 | - | 1 | 253 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MX dynamin-like GTPase 1 | Mx1 |
AI893580, Mx, Mx-1 |
-3.6kb | Sense | Chr. 16 - 97447035 - 97462906 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 3, member B |
Fam3b |
2-21, 9030624C24Rik, D16Jhu19e, ORF9, Pander |
4.1kb | Sense | Chr. 16 - 97471086 - 97504936 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2210009P08Rik
Genomic Map