Assay Details
| Gene Symbol: Vsig10l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290316.1 | NP_001277245.1 | 10 | 3234 | 64 | |
| GenBank mRNA | AK008908.1 | - | 1 | 748 | 64 | |
| EH095446.1 | - | 1 | 177 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
electron transferring flavoprotein, beta polypeptide |
Etfb |
0610009I16Rik, 2810441H06Rik |
-13.3kb | Sense | Chr. 7 - 43444072 - 43457800 on NCBI Build GRCm38 |
| Downstream | IgLON family member 5 | Iglon5 |
A230106M20Rik, Gm1431 |
894.0 | Anti-sense | Chr. 7 - 43472907 - 43490057 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vsig10l
Genomic Map