Assay Details
| Gene Symbol: Gpank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009290.1 | - | 288 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DNA segment, Chr 17, human D6S53E |
D17H6S53E |
G4, NG34 |
Anti-sense | Chr. 17 - 35126402 - 35128855 on NCBI Build GRCm38 | |
| Downstream | apolipoprotein M | Apom |
1190010O19Rik, G3a, NG20 |
2.6kb | Sense | Chr. 17 - 35128997 - 35131752 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gpank1
Genomic Map