Assay Details
| Gene Symbol: D730044K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021338.1 | - | 1 | 48 | 64 | |
| AK029389.1 | - | 1 | 679 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L30 | Rpl30 | -335.0 | Anti-sense | Chr. 15 - 34440505 - 34443276 on NCBI Build GRCm38 | |
| Downstream | glutamate rich 5 | Erich5 | 8.8kb | Sense | Chr. 15 - 34453312 - 34473892 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D730044K07Rik
Genomic Map