Assay Details
| Gene Symbol: Lgi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK031996.1 | - | 1 - 2 | 222 | 99 | |
| AK035873.1 | - | 1 - 2 | 290 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FRA10AC1 homolog (human) | Fra10ac1 |
5730455O13Rik, AI448293 |
-40.3kb | Anti-sense | Chr. 19 - 38188477 - 38224513 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 32960 |
Gm32960 | 79.6kb | Sense | Chr. 19 - 38348636 - 38350079 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Lgi1
Genomic Map