Assay Details
| Gene Symbol: 5830407E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037541.1 | - | 1 | 1354 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho GTPase activating protein 15 | Arhgap15 | 5830480G12Rik | Sense | Chr. 2 - 43748783 - 44395953 on NCBI Build GRCm38 | |
| Downstream |
Rho GTPase activating protein 15, opposite strand |
Arhgap15os | A430092G05Rik | 63.9kb | Anti-sense | Chr. 2 - 44059238 - 44065324 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5830407E08Rik
Genomic Map