Assay Details
| Gene Symbol: Xrcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038105.1 | - | 1 | 1942 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4117 | Gm4117 | -50.7kb | Anti-sense | Chr. 13 - 89734040 - 89798058 on NCBI Build GRCm38 | |
| Downstream | transmembrane protein 167 | Tmem167 |
0610041E09Rik, 5730424F14Rik, AU041184, AW537806, ENSMUSG00000060669, Gm10085, Tmem167a |
237.1kb | Anti-sense | Chr. 13 - 90089667 - 90114921 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Xrcc4
Genomic Map