Assay Details
| Gene Symbol: Ccr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040129.1 | - | 1 | 53 | 78 | |
| AK162996.1 | - | 1 | 119 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 17021 | Gm17021 | Anti-sense | Chr. 9 - 123767508 - 123772898 on NCBI Build GRCm38 | ||
| Downstream | FYVE and coiled-coil domain containing 1 | Fyco1 |
2810409M01Rik, Mem2, RUFY3, ZFYVE7 |
19.7kb | Anti-sense | Chr. 9 - 123789500 - 123851899 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccr9
Genomic Map