Assay Details
| Gene Symbol: B130055M24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041042.1 | - | 1 - 2 | 943 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 689 | Zfp689 |
4933416E05Rik, Znf689 |
Anti-sense | Chr. 7 - 127442136 - 127449158 on NCBI Build GRCm38 | |
| Downstream | proline rich 14 | Prr14 | BC006909 | 12.9kb | Sense | Chr. 7 - 127470096 - 127476767 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

B130055M24Rik
Genomic Map