Assay Details
| Gene Symbol: Nhsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041760.1 | - | 1 - 2 | 220 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 28A | Ccdc28a |
1700009P13Rik, AI480677 |
-172.6kb | Anti-sense | Chr. 10 - 18213685 - 18234991 on NCBI Build GRCm38 |
| Downstream | heme binding protein 2 | Hebp2 | SOUL | 131.6kb | Anti-sense | Chr. 10 - 18540123 - 18546076 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Nhsl1
Genomic Map