Assay Details
| Gene Symbol: H2afy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043531.1 | - | 505 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | paired-like homeodomain transcription factor 1 | Pitx1 |
Bft, P-OTX, Potx, Ptx1 |
-176.8kb | Anti-sense | Chr. 13 - 55825046 - 55836194 on NCBI Build GRCm38 |
| Downstream |
TRAF-interacting protein with forkhead-associated domain, family member B |
Tifab | 90.7kb | Anti-sense | Chr. 13 - 56173702 - 56179018 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

H2afy
Genomic Map