Assay Details
| Gene Symbol: Robo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034891.1 | - | 1 | 2118 | 102 | |
| AK046267.1 | - | 1 | 753 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930500H12 gene | 4930500H12Rik | -481.5kb | Anti-sense | Chr. 16 - 73072346 - 73410430 on NCBI Build GRCm38 | |
| Downstream | microRNA 691 | Mir691 |
Mirn691, mir-691, mmu-mir-691 |
449.0kb | Sense | Chr. 16 - 74341990 - 74342067 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Robo2
Genomic Map