Assay Details
| Gene Symbol: 4930435N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015328.1 | - | 1 | 1941 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 155, member A |
Fam155a |
6430500D16, AW121567, Tmem28 |
Anti-sense | Chr. 8 - 9206010 - 9773363 on NCBI Build GRCm38 | |
| Downstream |
ligase IV, DNA, ATP-dependent |
Lig4 |
5830471N16Rik, tiny |
508.8kb | Anti-sense | Chr. 8 - 9970020 - 9976323 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930435N07Rik
Genomic Map