Assay Details
| Gene Symbol: 4930480K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015601.1 | - | 1 | 950 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAS-like, estrogen-regulated, growth-inhibitor |
Rerg | Sense | Chr. 6 - 137054817 - 137170496 on NCBI Build GRCm38 | ||
| Downstream |
protein tyrosine phosphatase, receptor type, O |
Ptpro |
D28, GLEPP1, PTPBK, PTPROt, PTPU2, PTPoc, PTPphi, Ptpn15, R-PTP-O |
105.4kb | Anti-sense | Chr. 6 - 137252299 - 137464634 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930480K02Rik
Genomic Map