Assay Details
| Gene Symbol: 4930486N12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015636.1 | - | 1 | 451 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | otoancorin | Otoa | -95.3kb | Anti-sense | Chr. 7 - 121083438 - 121163470 on NCBI Build GRCm38 | |
| Downstream | heparan sulfate (glucosamine) 3-O-sulfotransferase 2 | Hs3st2 |
6430516N12Rik, A830061E14Rik, AW491345 |
132.2kb | Anti-sense | Chr. 7 - 121391610 - 121501768 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930486N12Rik
Genomic Map