Assay Details
Gene Symbol: 4930520E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015663.1 | - | 1 | 641 | 103 | |
AK015850.1 | - | 1 | 570 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | katanin p60 subunit A-like 2 | Katnal2 |
3110023G01Rik, 4933439B08Rik |
-16.2kb | Sense | Chr. 18 - 76977147 - 77048225 on NCBI Build GRCm38 |
Downstream | protein inhibitor of activated STAT 2 | Pias2 |
6330408K17Rik, AI462206, ARIP3, AU018068, Dib, Miz1, PIASxalpha, PIASxb, PIASxbeta, SIZ2 |
81.0 | Anti-sense | Chr. 18 - 77065208 - 77155710 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
