Assay Details
| Gene Symbol: Gm9806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015803.1 | - | 1 | 1244 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | vacuolar protein sorting 13A (yeast) | Vps13a |
4930425F11, 4930516E05Rik, 4930543C13Rik, 9930023P20, Chac, Chorein, D330038K10Rik, mKIAA0986 |
Anti-sense | Chr. 19 - 16612194 - 16781139 on NCBI Build GRCm38 | |
| Downstream | forkhead box B2 | Foxb2 | Fkh4 | 90.3kb | Anti-sense | Chr. 19 - 16872316 - 16873830 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm9806
Genomic Map