Assay Details
| Gene Symbol: 4930542N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016024.1 | - | 1 | 1049 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26519 |
Gm26519 | -6.3kb | Anti-sense | Chr. 5 - 105401226 - 105406964 on NCBI Build GRCm38 | |
| Downstream |
leucine rich repeat containing 8 family, member B |
Lrrc8b |
R75581, Ta-lrrp, mKIAA0231 |
105.0 | Anti-sense | Chr. 5 - 105415775 - 105490071 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930542N06Rik
Genomic Map