Assay Details
| Gene Symbol: 4930573C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016287.1 | - | 1 | 153 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
KH domain containing, RNA binding, signal transduction associated 3 |
Khdrbs3 |
Etle, SLM-2, Salp, Slm2, T-STAR |
-132.8kb | Anti-sense | Chr. 15 - 68928405 - 69101163 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930504C09 gene | 4930504C09Rik | 35.5kb | Anti-sense | Chr. 15 - 69270541 - 69274718 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930573C08Rik
Genomic Map