Assay Details
| Gene Symbol: 4930584E12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016348.1 | - | 1 | 854 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CWC15 homolog (S. cerevisiae) | Cwc15 |
0610040D20Rik, 2900052N06Rik, Ed1, c11orf5, mED1 |
-108.0kb | Anti-sense | Chr. 9 - 14500606 - 14510620 on NCBI Build GRCm38 |
| Downstream | predicted gene 7607 | Gm7607 | EG665374 | 43.5kb | Anti-sense | Chr. 9 - 14663020 - 14665100 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930584E12Rik
Genomic Map