Assay Details
| Gene Symbol: 4930588A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016363.1 | - | 3 - 4 | 595 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA C030004G16 gene | C030004G16Rik | -315.8kb | Sense | Chr. 18 - 42615618 - 42622739 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930572J10 gene | 4930572J10Rik | 135.6kb | Sense | Chr. 18 - 43074968 - 43087652 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930588A03Rik
Genomic Map