Assay Details
Gene Symbol: 4930595O18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016395.1 | - | 1 | 357 | 140 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
protein phosphatase 4, regulatory subunit 2 |
Ppp4r2 |
BE691708, C230060M08Rik |
-52.6kb | Anti-sense | Chr. 6 - 100833623 - 100869934 on NCBI Build GRCm38 |
Downstream |
predicted gene, 26911 |
Gm26911 | 136.0kb | Anti-sense | Chr. 6 - 101059133 - 101221620 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
