Assay Details
| Gene Symbol: 4932433N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016544.1 | - | 1 | 2248 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10910 | Gm10910 | ENSMUSG00000078079 | -97.3kb | Sense | Chr. 9 - 31280669 - 31281727 on NCBI Build GRCm38 |
| Downstream | nuclear factor related to kappa B binding protein | Nfrkb | A530090G11Rik | 4.4kb | Sense | Chr. 9 - 31386161 - 31421333 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4932433N03Rik
Genomic Map