Assay Details
| Gene Symbol: 4933412L11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016793.1 | - | 1 | 645 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5577 | Gm5577 | EG434064 | -9.0kb | Anti-sense | Chr. 6 - 87981683 - 87984180 on NCBI Build GRCm38 |
| Downstream |
RAB7, member RAS oncogene family |
Rab7 | Rab7a | 4.9kb | Sense | Chr. 6 - 87999106 - 88045270 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933412L11Rik
Genomic Map