Assay Details
| Gene Symbol: 4933428C20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016962.1 | - | 1 | 752 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/threonine kinase 33 | Stk33 |
4921505G21Rik, AW061264 |
-55.0kb | Anti-sense | Chr. 7 - 109279213 - 109439053 on NCBI Build GRCm38 |
| Downstream | ribosomal protein L27A | Rpl27a |
L27', L27A, L29 |
24.1kb | Sense | Chr. 7 - 109519195 - 109522369 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933428C20Rik
Genomic Map