Assay Details
| Gene Symbol: 4933440K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017135.1 | - | 1 | 1353 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | acyl-CoA thioesterase 12 | Acot12 |
1300004O04Rik, 4930449F15Rik, AV027244, Cach, mCACH-1 |
Anti-sense | Chr. 13 - 91741511 - 91786148 on NCBI Build GRCm38 | |
| Downstream |
zinc finger, CCHC domain containing 9 |
Zcchc9 | 1810019C21Rik | 14.2kb | Sense | Chr. 13 - 91796520 - 91807997 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933440K10Rik
Genomic Map