Assay Details
| Gene Symbol: 5033423O07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017189.1 | - | 1 | 751 | 126 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1110059E24 gene | 1110059E24Rik | -7.7kb | Anti-sense | Chr. 19 - 21597313 - 21652791 on NCBI Build GRCm38 | |
| Downstream | transmembrane protein 2 | Tmem2 | 115.9kb | Sense | Chr. 19 - 21777508 - 21858437 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5033423O07Rik
Genomic Map