Assay Details
| Gene Symbol: 5430416G10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017315.1 | - | 1 | 1453 | 75 | |
| BB086802.1 | - | 1 | 156 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4221 | Gm4221 | Sense | Chr. 18 - 54990246 - 55143754 on NCBI Build GRCm38 | ||
| Downstream | GRAM domain containing 3 | Gramd3 |
9030613F08Rik, 9130427A09Rik, AW061306, NS3TP2 |
1.3mb | Sense | Chr. 18 - 56387485 - 56503792 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5430416G10Rik
Genomic Map