Assay Details
Gene Symbol: Pus3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017552.1 | - | 1 | 1245 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 25 | Ddx25 |
AW047046, GRTH |
-1.0kb | Anti-sense | Chr. 9 - 35541848 - 35558470 on NCBI Build GRCm38 |
Downstream | prostate and testis expressed 2 | Pate2 |
Gm846, Pate-M, mANLP1 |
13.5kb | Sense | Chr. 9 - 35574309 - 35698689 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
