Assay Details
| Gene Symbol: 5730492I20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017720.1 | - | 1 | 1148 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SH3-domain binding protein 4 | Sh3bp4 |
AI594717, AW227605, BOG25 |
Sense | Chr. 1 - 89070462 - 89155084 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA C030007H22 gene | C030007H22Rik | 220.4kb | Anti-sense | Chr. 1 - 89360342 - 89406133 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5730492I20Rik
Genomic Map