Assay Details
| Gene Symbol: Gpkow | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017864.1 | - | 1 | 752 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prefoldin 5 | Pfdn5 |
1190001O17Rik, 1700010A06Rik, D15Ertd697e, Eig1, MM-1 |
Anti-sense | Chr. 15 - 102326116 - 102331490 on NCBI Build GRCm38 | |
| Downstream | WD repeat domain 45 | Wdr45 |
C79260, DXImx38e, JM5, Sfc19, WIPI-4, Wdrx1 |
23.9kb | Sense | Chr. X - 7721995 - 7728201 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpkow
Genomic Map