Assay Details
| Gene Symbol: 5830438M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017980.1 | - | 1 | 944 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger protein 168 | Rnf168 | 3110001H15Rik | -1.7kb | Sense | Chr. 16 - 32277461 - 32301442 on NCBI Build GRCm38 |
| Downstream | UBX domain protein 7 | Ubxn7 |
A630090P18, AI196514, Ubxd7, mKIAA0794 |
27.7kb | Sense | Chr. 16 - 32332184 - 32393750 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5830438M01Rik
Genomic Map