Assay Details
| Gene Symbol: 5830453K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018007.1 | - | 1 | 1353 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PC-esterase domain containing 1B | Pced1b | Fam113b | -38.2kb | Anti-sense | Chr. 15 - 97247080 - 97385696 on NCBI Build GRCm38 |
| Downstream | RNA polymerase II associated protein 3 | Rpap3 |
2310042P20Rik, D15Ertd682e |
249.5kb | Sense | Chr. 15 - 97675097 - 97705822 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5830453K13Rik
Genomic Map