Assay Details
| Gene Symbol: 5830454J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018012.1 | - | 1 | 449 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase 14 | Mapk14 |
CSBP2, Crk1, Csbp1, Mxi2, PRKM14, PRKM15, p38, p38-alpha, p38MAPK, p38a, p38alpha |
Sense | Chr. 17 - 28691342 - 28748405 on NCBI Build GRCm38 | |
| Downstream | mitogen-activated protein kinase 13 | Mapk13 |
SAPK4, Serk4 |
73.6kb | Sense | Chr. 17 - 28769317 - 28778704 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5830454J16Rik
Genomic Map