Assay Details
| Gene Symbol: 6330531I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI643441.1 | - | 1 | 76 | ||
| AK018224.1 | - | 1 | 1450 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 1587 | Gm1587 | -21.7kb | Sense | Chr. 14 - 77793945 - 77798968 on NCBI Build GRCm38 | |
| Downstream |
DnaJ (Hsp40) homolog, subfamily C, member 15 |
Dnajc15 |
1110003P16Rik, Dnajd1 |
3.9kb | Sense | Chr. 14 - 77826217 - 77874917 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6330531I01Rik
Genomic Map