Assay Details
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018352.1 | - | 1 | 1950 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13363 | Gm13363 | OTTMUSG00000011595 | -762.5kb | Sense | Chr. 2 - 19992266 - 19995055 on NCBI Build GRCm38 |
| Downstream | Rho GTPase activating protein 21 | Arhgap21 |
5530401C11Rik, AA416458, ARHGAP10 |
88.3kb | Anti-sense | Chr. 2 - 20847919 - 20968638 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Etl4
Genomic Map