Assay Details
| Gene Symbol: 9030418E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018517.1 | - | 1 | 850 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | low density lipoprotein-related protein 1B (deleted in tumors) | Lrp1b |
9630004P12Rik, LRP-DIT |
Sense | Chr. 2 - 40595290 - 42653631 on NCBI Build GRCm38 | |
| Downstream | microRNA 6336 | Mir6336 |
Gm27584, mmu-mir-6336 |
308.7kb | Sense | Chr. 2 - 42447439 - 42447568 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9030418E23Rik
Genomic Map