Assay Details
| Gene Symbol: D430040D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052512.1 | - | 1 | 1857 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C |
Sema4c |
AI426163, M-Sema F, Semacl1, Semaf, Semai, sema I |
Anti-sense | Chr. 1 - 36548640 - 36558381 on NCBI Build GRCm38 | |
| Downstream |
family with sequence similarity 178, member B |
Fam178b |
1700024G10Rik, Gm990 |
3.0kb | Anti-sense | Chr. 1 - 36562696 - 36683190 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D430040D24Rik
Genomic Map