Assay Details
Gene Symbol: Med26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK054023.1 | - | 1548 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC102639683 | LOC102639683 | Sense | Chr. 8 - 72492731 - 72504592 on NCBI Build GRCm38 | ||
Downstream | small integral membrane protein 7 | Smim7 | 9130011J15Rik | 70.3kb | Anti-sense | Chr. 8 - 72565198 - 72571048 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
