Assay Details
| Gene Symbol: Pard3bos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015421.1 | - | 440 | 148 | ||
| AK076674.1 | - | 2 - 3 | 518 | 148 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9530026F06 gene | 9530026F06Rik | -390.8kb | Anti-sense | Chr. 1 - 61378432 - 61395904 on NCBI Build GRCm38 | |
| Downstream | neuropilin 2 | Nrp2 |
1110048P06Rik, Np-2, Np2, Npn-2, Npn2 |
854.8kb | Anti-sense | Chr. 1 - 62703317 - 62818692 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Pard3bos1
Genomic Map