Assay Details
| Gene Symbol: Tmem30a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078136.1 | - | 1 | 1248 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytochrome c oxidase subunit VIIa 2 | Cox7a2 |
COX7AL, Cox7a3, CoxVIIa-L |
-8.4kb | Sense | Chr. 9 - 79755241 - 79759853 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930429F24 gene | 4930429F24Rik | 23.7kb | Anti-sense | Chr. 9 - 79793641 - 79803235 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmem30a
Genomic Map