Assay Details
| Gene Symbol: A730063M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040660.1 | 2 | 227 | 72 | ||
| GenBank mRNA | AA789631.1 | - | 2 | 233 | 72 | |
| AK078153.1 | - | 1 | 544 | 72 | ||
| AK080508.1 | - | 1 | 627 | 72 | ||
| BX529939.1 | - | 2 | 228 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetraspanin 31 | Tspan31 |
2700085A14Rik, AW742554, Sas |
-196.0 | Anti-sense | Chr. 10 - 127067290 - 127070261 on NCBI Build GRCm38 |
| Downstream |
ArfGAP with GTPase domain, ankyrin repeat and PH domain 2 |
Agap2 |
AGAP-2, Centg1, PIKE, cnt-g1, mKIAA0167 |
4.2kb | Sense | Chr. 10 - 127075258 - 127093173 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A730063M14Rik
Genomic Map