Assay Details
| Gene Symbol: Sema6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078490.1 | - | 1 | 3454 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | COMM domain containing 10 | Commd10 |
2310003A05Rik, AU019438, DRWMS2 |
-193.7kb | Anti-sense | Chr. 18 - 46958785 - 47087995 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 9130209A04 gene | 9130209A04Rik | 167.5kb | Anti-sense | Chr. 18 - 47452668 - 47470868 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sema6a
Genomic Map