Assay Details
| Gene Symbol: Arhgap15os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040622.1 | 1 - 2 | 85 | 103 | ||
| GenBank mRNA | AK078625.1 | - | 1 - 2 | 86 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho GTPase activating protein 15 | Arhgap15 | 5830480G12Rik | Anti-sense | Chr. 2 - 43748783 - 44395953 on NCBI Build GRCm38 | |
| Downstream | glycosyltransferase-like domain containing 1 | Gtdc1 | E330008O22Rik | 499.1kb | Sense | Chr. 2 - 44564412 - 44927918 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap15os
Genomic Map